ContactUsLinkComponent
Murine IL-22 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombina nt murine IL-22 in a sandwich ELISA format. Using the ELISA protocol described below, the recommended microplates, r 1 * 200 Tests
  PEPR900-M257
Murine IL-22 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombina nt murine IL-22 in a sandwich ELISA format. Using the ELISA protocol described below, the recommended microplates, r 1 * 200 Tests
  PEPR900-M257
Frequently Bought Together