ContactUsLinkComponent
Murine MIP-2 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombin ant mMIP-2 in a sandwich ELISA format. Using the ELISA protocol described below, the recommended microplates, reagent 1 * 200 Tests
Catalog # PEPR900-M152
Supplier:  PeproTech, Inc.
Murine MIP-2 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombin ant mMIP-2 in a sandwich ELISA format. Using the ELISA protocol described below, the recommended microplates, reagent 1 * 200 Tests
Catalog # PEPR900-M152
Supplier:  PeproTech, Inc.
Frequently Bought Together