Recombinant Human TGF-beta1 (HEK293 derived) 1 * 50 µG
Catalog # PEPR100-21-50UG
Supplier: PeproTech, Inc.
Recombinant Human TGF-beta1 (HEK293 derived) 1 * 50 µG
Catalog # PEPR100-21-50UG
Supplier: PeproTech, Inc.
Frequently Bought Together
Nalgene®, Biotainer® Bottles
Supplier: Thermo Fisher Scientific
Media bottles, square, sterile, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
Microplates, 96-well
Supplier: Greiner Bio-One
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Racks for culture tubes, stainless steel
Supplier: BELLCO GLASS
Dosing spoon and measuring spoon set
Supplier: BURKLE
Lab Consumables
Supplier: Copan
Petri dish racks, stainless steel, PetriPile®
Supplier: INTERSCIENCE
MICROTITER PLATE U FORM STERILE 1 * 50 items
Supplier: DELTALAB
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
Axygen® Microcentrifuge Screw Caps
Supplier: Corning
Collagen I-coated microplates, Corning® BioCoat®
Supplier: Corning
Storage plates, 96-well, MASTERBLOCK®
Supplier: Greiner Bio-One