Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Seed testing papers, Munktell

37/N, 580×580 mm
  BINZ213030
 :   Ahlstrom
Seed testing papers, Munktell
  BINZ213030
 :  Ahlstrom
Seed testing papers, Munktell
Seed testing papers, Munktell

37/N, 580×580 mm

 
SFr. 344.00
   
1 * 100 ST
 

Seed Testing Papers are widely used by seed testing institutes and others for germination capacity testing and for health tests e.g. Jacobsen (Copenhagen tank), on top of paper and osmosis test.

 
  • Grade:
    37/N
  • Seq:
    02
  • Size:
    580×580 mm
  • Cat. No.:
    BINZ213030
  • Capillary rise:
    -
  • Ash content:
    -
  • Weight:
    135 g/m²