Tanks, rectangular, LDPE, LaboPlast®
90 L, Seamless containers, rectangular shaped, - mm
Catalog # BURK3440-0906
Supplier: BURKLE
Jump to:
Product Details & Documents
These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.
- Linear LDPE, transparent
- Rupture-proof
Specifications
- Capacity:90 L
- Description:Seamless containers, rectangular shaped
- Seq:07
- W×D×H:- mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Container thread ¾" for cylindrical containersContainer thread ¾" for cylindrical containersBURK3440-0034Log in for availability1 * 1 ST - SFr. 202.00