Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Detergent solutions, Surfact-Amps™

Detergent solutions, Surfact-Amps™

Triton X-100 Surfact-Amps™ detergent solution, 50 mL
Catalog # PIER85111
Detergent solutions, Surfact-Amps™
Catalog # PIER85111
CAS Number:  
Detergent solutions, Surfact-Amps™
Detergent solutions, Surfact-Amps™

Triton X-100 Surfact-Amps™ detergent solution, 50 mL

CAS Number:  
Price
SFr. 281.00
Log in for availability
1 * 50 mL
Product Details & Documents

Surfact-Amps™ detergents are ready-to-use, diluted solutions of purified detergent that can be used in routine and high-demand protein research methods and molecular biology techniques. These detergents are useful as a component of cell lysis buffers or other solutions intended to extract and solubilise proteins, improve chromatography, prepare yeast spheroplasts, and reduce background in ELISA or Western blot methods.

  • Clear and pure 10% detergent solutions with <1.0 μeq/ml peroxides and carbonyl content
  • Ampoules are packed under nitrogen for maximum shelf life and are pre-scored for easy opening
  • Easy to pipette and accurately dispense
  • Packaging eliminates the accumulation of peroxides and degradation products

Detergents are a class of molecules whose unique properties enable manipulation (disruption or formation) of hydrophobichydrophilic interactions among molecules in biological samples. In biological research, detergents are used to lyse cells (release soluble proteins), solubilise membrane proteins and lipids, control protein crystallisation and prevent non specific binding in affinity purification and immunoassay procedures. They are often used as an additive in protein gel electrophoresis.

Documents

Specifications

Specifications

  • Pk:
    50 mL
  • Description:
    Triton X-100 Surfact-Amps™ detergent solution

Specifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.