Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Detergent solutions, Surfact-Amps™

Tween™ 20 Surfact-Amps™ detergent solution, 50 mL
Catalog # PIER85113
Detergent solutions, Surfact-Amps™
Catalog # PIER85113
CAS Number:  
Detergent solutions, Surfact-Amps™
Detergent solutions, Surfact-Amps™

Tween™ 20 Surfact-Amps™ detergent solution, 50 mL

CAS Number:  
Price
SFr. 285.00
Log in for availability
1 * 50 mL
Product Details & Documents

Surfact-Amps™ detergents are ready-to-use, diluted solutions of purified detergent that can be used in routine and high-demand protein research methods and molecular biology techniques. These detergents are useful as a component of cell lysis buffers or other solutions intended to extract and solubilise proteins, improve chromatography, prepare yeast spheroplasts, and reduce background in ELISA or Western blot methods.

  • Clear and pure 10% detergent solutions with <1.0 μeq/ml peroxides and carbonyl content
  • Ampoules are packed under nitrogen for maximum shelf life and are pre-scored for easy opening
  • Easy to pipette and accurately dispense
  • Packaging eliminates the accumulation of peroxides and degradation products

Detergents are a class of molecules whose unique properties enable manipulation (disruption or formation) of hydrophobichydrophilic interactions among molecules in biological samples. In biological research, detergents are used to lyse cells (release soluble proteins), solubilise membrane proteins and lipids, control protein crystallisation and prevent non specific binding in affinity purification and immunoassay procedures. They are often used as an additive in protein gel electrophoresis.

 

Specifications

Specifications

  • Pk:
    50 mL
  • Description:
    Tween™ 20 Surfact-Amps™ detergent solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.