StartingBlock™ Blocking Buffer
StartingBlock™ (PBS), blocking buffer, 100 ml, 100 ml
PIER37578
These StartingBlock™ blocking buffers are PBS and TBS formulations of a proprietary protein that provides exceptional performance and broad compatibility with membrane (Western blot) and 96-well plate (ELISA) assays.
- Compatible with many detection systems (Western blot, ELISA and IHC)
- Short blocking times
- High signal:noise ratio
- Strip and reprobe without reblocking
- Available ready to use 1X formulations in PBS and TBS
- For research use only
- Not for use in diagnostic procedures
It provides broad compatibility with a wide range of antibodies, antibody combinations, and other protein probing and assay systems. StartingBlock blocking buffer is available in PBS and TBS, with and without Tween 20.
- Pk:100 ml
- Description:StartingBlock™ (PBS), blocking buffer, 100 ml
- Cat. No.:PIER37578
- Physical form:Liquid
- Concentration:1X
- Storage buffer:PBS
- For use with:Western blot, ELISA, Immunohistochemistry (IHC)
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



