Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

StartingBlock™ Blocking Buffer

StartingBlock™ (TBS), blocking buffer, 100 ml, 100 ml
  PIER37579
StartingBlock™ Blocking Buffer
  PIER37579
 :  
StartingBlock™ Blocking Buffer
StartingBlock™ Blocking Buffer

StartingBlock™ (TBS), blocking buffer, 100 ml, 100 ml

 :  
 
89.20 SFr.
   
1 * 100 mL
 

These StartingBlock™ blocking buffers are PBS and TBS formulations of a proprietary protein that provides exceptional performance and broad compatibility with membrane (Western blot) and 96-well plate (ELISA) assays.

  • Compatible with many detection systems (Western blot, ELISA and IHC)
  • Short blocking times
  • High signal:noise ratio
  • Strip and reprobe without reblocking
  • Available ready to use 1X formulations in PBS and TBS
  • For research use only
  • Not for use in diagnostic procedures

It provides broad compatibility with a wide range of antibodies, antibody combinations, and other protein probing and assay systems. StartingBlock blocking buffer is available in PBS and TBS, with and without Tween 20.

 

 

 

  • Pk:
    100 ml
  • Description:
    StartingBlock™ (TBS), blocking buffer, 100 ml
  • Cat. No.:
    PIER37579
  • Physical form:
    Liquid
  • Concentration:
    1X
  • Storage buffer:
    TBS
  • For use with:
    Western blot, ELISA, Immunohistochemistry (IHC)