ContactUsLinkComponent
Each v-21+k wear metals standard 500 µg/g, hydrocarbon oil, 100 g includes the following elements: ag, al, ba, b, cd, ca, cr, cu, fe , k, mg, mn, mo, na, ni, p, pb, si, sn, ti, v, zn. concentration: 500 µg/g, matrix: hydrocarbon oil and volume: 100 g 1 * 1 items
Catalog # PERCN9308365
Supplier:  PerkinElmer
Each v-21+k wear metals standard 500 µg/g, hydrocarbon oil, 100 g includes the following elements: ag, al, ba, b, cd, ca, cr, cu, fe , k, mg, mn, mo, na, ni, p, pb, si, sn, ti, v, zn. concentration: 500 µg/g, matrix: hydrocarbon oil and volume: 100 g 1 * 1 items
Catalog # PERCN9308365
Supplier:  PerkinElmer
Frequently Bought Together