Ultra clean filter bundle of 3 for gc. includes (1) oxygen/moisture/hydrocarbon trap and (2) hydrocarbon/ moisture traps. its featur es and benefits include - ultra-high capacity high-purity output ensures 99.9999% pure gas quick connect fittings f 1 * 1 items
Catalog # PERCN9306826
Supplier: PerkinElmer
Frequently Bought Together