Closures for headspace vials
Red PTFE coated rubber cap/septa/spring, pre-assembled
Catalog # PERCN9302978
Supplier: PerkinElmer
Jump to:
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Specifications
- Description:Red PTFE coated rubber cap/septa/spring, pre-assembled
- Seq:22