ContactUsLinkComponent
Used for hydrocarbon removal. use a hydrocarbon trap if your gas has a potential source of hydrocarbon contaminants or if you suspec t you are observing carrier gas ghost peaks. install the hydrocarbon trap after the moisture trap, to prevent moistur 1 * 1 items
Catalog # PERCN9306816
Supplier:  PerkinElmer
Used for hydrocarbon removal. use a hydrocarbon trap if your gas has a potential source of hydrocarbon contaminants or if you suspec t you are observing carrier gas ghost peaks. install the hydrocarbon trap after the moisture trap, to prevent moistur 1 * 1 items
Catalog # PERCN9306816
Supplier:  PerkinElmer
Frequently Bought Together