ContactUsLinkComponent
footpump, stainless steel for dissolvents for drums up to 220 l. stiff outlet elbow with stopcock, stainless steel/ptfe, max. immers ion: 950 mm, dip tube adjustable to drum depth. extraction capacity: 30 (l/min) r 2 fine thread 1 * 1 items
Catalog # MSGA16841
Supplier:  asecos
footpump, stainless steel for dissolvents for drums up to 220 l. stiff outlet elbow with stopcock, stainless steel/ptfe, max. immers ion: 950 mm, dip tube adjustable to drum depth. extraction capacity: 30 (l/min) r 2 fine thread 1 * 1 items
Catalog # MSGA16841
Supplier:  asecos
Frequently Bought Together