Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Safety dispensing canisters

Safety dispensing canisters

20 l, 175×345×470 mm
Catalog # MSGA16781
Supplier:   asecos
Safety dispensing canisters
Catalog # MSGA16781
Supplier:  asecos
Safety dispensing canisters
Safety dispensing canisters

20 l, 175×345×470 mm

Price
SFr. 712.00
Log in for availability
1 * 1 ST
Product Details & Documents

Canister, made of stainless steel, with tap and separate ventilation.

  • Manual pressure
  • Vacuum balancing
  • Self-closing tap
Specifications
  • Capacity:
    20 l
  • Seq:
    03
  • W×D×H:
    175×345×470 mm
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Nozzle for filling canisters
    Nozzle for filling canisters
    MSGA16796
    Log in for availability
    1 * 1 ST - SFr. 189.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.