ContactUsLinkComponent
VWR 1.5ml loop screw cap tubes and caps. rnase/dnase free, non-pyrogenic. autoclavable. clear cap with silicone seal. 500 caps an d 500 tubes per resealable package. sterile. gamma irradiated. 1 * 500 items
Catalog # 211-0439
Supplier:  VWR Collection
undefined
VWR 1.5ml loop screw cap tubes and caps. rnase/dnase free, non-pyrogenic. autoclavable. clear cap with silicone seal. 500 caps an d 500 tubes per resealable package. sterile. gamma irradiated. 1 * 500 items
Catalog # 211-0439
Supplier:  VWR Collection
Frequently Bought Together