ContactUsLinkComponent
Histology: Sense Organs, 10 microscope slides - 1. Eye of cat, posterior part with retina, sagittal l.s. 2. Eye of cat, anterior par t with iris, ciliary body, cornea, sagittal l.s. 3. Developing eyes in t.s. of head from guinea pig embryo. 4. Retina 1 * 1 items
Catalog # LIED72280
Supplier:  LIEDER JOHANNES
Histology: Sense Organs, 10 microscope slides - 1. Eye of cat, posterior part with retina, sagittal l.s. 2. Eye of cat, anterior par t with iris, ciliary body, cornea, sagittal l.s. 3. Developing eyes in t.s. of head from guinea pig embryo. 4. Retina 1 * 1 items
Catalog # LIED72280
Supplier:  LIEDER JOHANNES
Frequently Bought Together