ContactUsLinkComponent
Human Histology, Large Set Part II. - 50 Microscope Slides - 1. Soft palate, human t.s. 2. Adipose tissue, human, sec. stained for f at 3. White fibrous cartilage, human intervertebral disc, sec. 4. Striated (skeletal) muscle, human t.s. 5. Spongy (c 1 * 1 items
Catalog # LIED72000
Supplier:  LIEDER JOHANNES
Human Histology, Large Set Part II. - 50 Microscope Slides - 1. Soft palate, human t.s. 2. Adipose tissue, human, sec. stained for f at 3. White fibrous cartilage, human intervertebral disc, sec. 4. Striated (skeletal) muscle, human t.s. 5. Spongy (c 1 * 1 items
Catalog # LIED72000
Supplier:  LIEDER JOHANNES
Frequently Bought Together