ContactUsLinkComponent
kit, mycoplasma pneumoniae igg elisa kit, mycoplasma pneumoniae igg elisa kit isintended for the detection of igg antibodies to mycoplasma pneumoniae in human serum or plasma. 1 * 1 KIT
Catalog # ABNOKA2260
Supplier:  Abnova
kit, mycoplasma pneumoniae igg elisa kit, mycoplasma pneumoniae igg elisa kit isintended for the detection of igg antibodies to mycoplasma pneumoniae in human serum or plasma. 1 * 1 KIT
Catalog # ABNOKA2260
Supplier:  Abnova