ContactUsLinkComponent
dna probe, fish probe, pdgfra/fip1l1 3-color rearrangement fish probe, labeled fish probes for identification of gene translocation using fluorescent in situ hybridization technique. target= _blank >technology) 1 * 200 µl
Catalog # ABNOFT0008
Supplier:  Abnova
dna probe, fish probe, pdgfra/fip1l1 3-color rearrangement fish probe, labeled fish probes for identification of gene translocation using fluorescent in situ hybridization technique. target= _blank >technology) 1 * 200 µl
Catalog # ABNOFT0008
Supplier:  Abnova
Frequently Bought Together