Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Homogenisers, behrotest®

Homogenisers, behrotest®

Homogeniser, 15 litre capacity, QMR 15
Catalog # 331-3310
Supplier:   BEHR
Homogenisers, behrotest®
Catalog # 331-3310
Supplier:  BEHR
Homogenisers, behrotest®
Homogenisers, behrotest®

Homogeniser, 15 litre capacity, QMR 15

Price
SFr. 4’000.00
Log in for availability
1 * 1 ST
Product Details & Documents

Homogenisers for pretreatment, sample splitting and homogenisation of heterogeneous water samples in accordance with German waste water quality control standards.

  • The sample vessel is protected against damage by the homogeniser frame, which has two handles for easy carrying
  • Sample vessel volumes from 5 to 25 litre permit multiple sample collection (mixed samples)
  • Graduated for easy measurement of sample volume
  • Adjustable-speed magnetic stirrer
  • Withdrawl of desired volume of homogenized sample by means of a PTFE stopcock
  • Can be operated from a 12 V auto battery using an optional voltage converter
Specifications
  • Description:
    Homogeniser, 15 litre capacity, QMR 15
  • Seq:
    02
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Behrotest glass vessel, 15 l
    Behrotest glass vessel, 15 l
    331-3312
    Log in for availability
    1 * 1 ST - SFr. 1’270.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.