Blitz Bionsensor binds specifically to the Fc portion of human iggs and other proteins containing a human Fc region. applications i nclude quantitation of human igg and proteins containing the human Fc region in cell line development, clone selectio 1 * 1 items
Catalog # 733-2136
Supplier: PALL FORTEBIO
Frequently Bought Together