Orders LinkContactUsLinkComponent
Multi-panel drug tests
Multi-panel drug tests
Multi-panel drug tests, (AMP/mAMP/BAR/BZO/COC/MDMA/MTD/OPI/TCA/THC)
Catalog # SUREG10DS2
Supplier:   SURESCREEN
Multi-panel drug tests
Catalog # SUREG10DS2
Supplier:  SURESCREEN
Multi-panel drug tests
Multi-panel drug tests
Multi-panel drug tests, (AMP/mAMP/BAR/BZO/COC/MDMA/MTD/OPI/TCA/THC)
Price
SFr. 291.00
Log in for availability
1 * 25 ST
Product Details & Documents
These multipanel drugs tests can detect up to 12 drugs simultaneously.
  • Clear results in less than three minutes
  • Great accuracy, wide range of tests available
  • For detecting recent drug use approximately 1 to 8 days
Suitable for on-site screening, safety critical situations, random screening, for cause screening, and monitoring rehabilitation.
AMP: Amphetamines ; BAR: Barbiturates ; BUP: Buprenorphine ; BZO: Benzodiazepines ; CLON: Clonezepam ; COC: Cocaine ; EDDP: Methadone Metabolite ; FEN: Fentanyl ; KET: Ketamine ; MAMP: Methamphetamines ; MDMA: Ecstasy ; MOP: Morphine ; MTD: Methadone ; OPI: Opiates ; OXY: Oxycodone ; PCP: Phencyclidine ; PPX: Propoxyphene ; TCA: Tricyclic Antidepressants ; TRA: Tramadol ; THC: Cannabis.
Certifications: ISO 9001, ISO 13485. IVD in accordance with 98/79/EC.
Packaging: Individually wrapped in a foil pouch, which ensures that the high-quality of the product is retained throughout the shelf-life.
Specifications
  • Description:
    Multi-panel drug tests
  • Seq:
    033
  • Tests:
    (AMP/mAMP/BAR/BZO/COC/MDMA/MTD/OPI/TCA/THC)