Order Entry
Switzerland
Orders LinkContactUsLinkComponent
VWR®, Cell Holders

VWR®, Cell Holders

Cell holder (sample), 1 position for 10 mm standard cells
Catalog # 634-0777
Supplier:   VWR Collection
VWR®, Cell Holders
Catalog # 634-0777
Supplier:  VWR Collection
VWR®, Cell Holders
VWR®, Cell Holders

Cell holder (sample), 1 position for 10 mm standard cells

Price
SFr. 142.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Cell holder (sample), 1 position for 10 mm standard cells
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.