D-(-)-Serine
Glass bottle for solids, 100 g
Katalognr. ACTIAS21703G100
Marketsource
Anbieter: ACTIVATE SCIENTIFIC
D-(-)-Serine
Katalognr. ACTIAS21703G100
Anbieter: ACTIVATE SCIENTIFIC
Marketsource
CAS Nummer: 312-84-5
Direkt zu
Product Details & Documents
Synonyme: (R)-(-)-Serine, H-D-Ser-OH
- CAS Nummer:312-84-5
- Formel:C₃H₇NO₃
- MDL-Nummer:MFCD00004269
- Molekulargewicht:105.09 g/mol
- Merck-Index:12,08604
- Schmelzender Platingehalt:235 °C
- EINECS:206-229-4
Technische Daten
Technische Daten
- Pack type:Glass bottle for solids
- Pk:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



