Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Inserts

Inserts

Insert 1/1 E 127 for 44 flasks 100 ml, PG 8535/36, PG 8582, PG 8582 CD, PG 8592, 445×445×102 mm
Catalog # MIEL3808530
Inserts
Catalog # MIEL3808530
Supplier:  
Inserts
Inserts

Insert 1/1 E 127 for 44 flasks 100 ml, PG 8535/36, PG 8582, PG 8582 CD, PG 8592, 445×445×102 mm

Discontinued
Product Details & Documents
Specifications
  • Description:
    Insert 1/1 E 127 for 44 flasks 100 ml
  • Seq:
    45
  • W×D×H:
    445×445×102 mm
  • For:
    PG 8535/36, PG 8582, PG 8582 CD, PG 8592
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.