2',3'-Dimethoxyacetophenone
Glass bottle for solids, 1 g
Catalog # ACTIAS36777G1
Marketsource
Supplier: ACTIVATE SCIENTIFIC
2',3'-Dimethoxyacetophenone
Catalog # ACTIAS36777G1
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 38480-94-3
Jump to:
Product Details & Documents
- CAS Number:38480-94-3
- Formula:C₁₀H₁₂O₃
- MDL Number:MFCD03424412
- Molecular Weight:180.2005 g/mol
- EINECS:214-468-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



