5-Bromopicolinamide
Glass bottle for solids, 5 g
Catalog # ACTIAS35418G5
Marketsource
Supplier: ACTIVATE SCIENTIFIC
5-Bromopicolinamide
Catalog # ACTIAS35418G5
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 90145-48-5
Jump to:
Product Details & Documents
Synonyms: 5-Bromopyridine-2-carboxamide
- CAS Number:90145-48-5
- Formula:C₆H₅BrN₂O
- MDL Number:MFCD04066715
- Molecular Weight:201.0200 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



