Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

5-Bromopicolinamide

Glass bottle for solids, 1 g
Catalog # ACTIAS35418G1
Marketsource
Supplier:   ACTIVATE SCIENTIFIC
5-Bromopicolinamide
Catalog # ACTIAS35418G1
Marketsource
CAS Number:  90145-48-5
5-Bromopicolinamide

Glass bottle for solids, 1 g

CAS Number:  90145-48-5
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  5-Bromopyridine-2-carboxamide
  • CAS Number:90145-48-5
  • Formula:C₆H₅BrN₂O
  • MDL Number:MFCD04066715
  • Molecular Weight:201.0200 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications