2-Fluoro-6-hydroxybenzonitrile
Glass bottle for solids, 5 g
Catalog # ACTIAS31471G5
Marketsource
Supplier: ACTIVATE SCIENTIFIC
2-Fluoro-6-hydroxybenzonitrile
Catalog # ACTIAS31471G5
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 140675-43-0
Jump to:
Product Details & Documents
- CAS Number:140675-43-0
- Formula:C₇H₄FNO
- MDL Number:MFCD03428592
- Molecular Weight:137.11 g/mol
- UN:3439
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



