2-Fluoro-6-hydroxybenzonitrile
Glass bottle for solids, 1 g
Catalog # ACTIAS31471G1
Marketsource
Supplier: ACTIVATE SCIENTIFIC
2-Fluoro-6-hydroxybenzonitrile
Catalog # ACTIAS31471G1
Supplier: ACTIVATE SCIENTIFIC
Marketsource
CAS Number: 140675-43-0
Jump to:
Product Details & Documents
- CAS Number:140675-43-0
- Formula:C₇H₄FNO
- MDL Number:MFCD03428592
- UN:3439
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



