Order Entry
Switzerland
Orders LinkContactUsLinkComponent
2,6-Dichlorophenethylamine

2,6-Dichlorophenethylamine

Glass bottle for solids, 5 g
Catalog # ACTIAS24939G5
Marketsource
Supplier:   ACTIVATE SCIENTIFIC
2,6-Dichlorophenethylamine
Catalog # ACTIAS24939G5
Marketsource
CAS Number:  14573-23-0
2,6-Dichlorophenethylamine

Glass bottle for solids, 5 g

CAS Number:  14573-23-0
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2-(2-Aminoethyl)-1,3-dichlorobenzene, 1-Amino-2-(2,6-dichlorophenyl)ethane, 2-(2,6-Dichlorophenyl)ethylamine, 2,6-Dichlorophenethylamine
  • CAS Number:14573-23-0
  • Formula:C₈H₉Cl₂N
  • MDL Number:MFCD00060612
  • Molecular Weight:190.07 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications