Accessories
Paper rolls APH 591, for PRT 100, 5 off, width 76 mm, Glassware washing units
Katalognr. 141-0640
Anbieter: MIELE
Direkt zu
Product Details & Documents
- For lower and upper baskets
- With 7 x 7 mm perforations
- Stainless steel
Technische Daten
- Description:Paper rolls APH 591, for PRT 100, 5 off, width 76 mm
- Seq:26
- For:Glassware washing units
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



