Process documentaion
APH 305 Extension cable 10 m, for the extension of connection cable type 1 and 2, All glassware washers
Catalog # 141-0621
Supplier: MIELE
Jump to:
Product Details & Documents
Specifications
- Description:APH 305 Extension cable 10 m, for the extension of connection cable type 1 and 2
- Seq:17
- For:All glassware washers
- Cat. No.:141-0621
- W×D×H:173×479×72 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



