Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Process documentaion

Process documentaion

APH 409 Crossover network cable for direct connection to PC, length 5 m (type 9), G 7823 , G 7824, G 7825, G 7826, G 7831, PG 8581, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8591, PG 8592, PG 8593
Catalog # 141-0626
Supplier:   MIELE
Process documentaion
Catalog # 141-0626
Supplier:  MIELE
Process documentaion
Process documentaion

APH 409 Crossover network cable for direct connection to PC, length 5 m (type 9), G 7823 , G 7824, G 7825, G 7826, G 7831, PG 8581, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8591, PG 8592, PG 8593

Price
SFr. 41.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    APH 409 Crossover network cable for direct connection to PC, length 5 m (type 9)
  • Seq:
    21
  • For:
    G 7823 , G 7824, G 7825, G 7826, G 7831, PG 8581, PG 8582, PG 8582 CD, PG 8583, PG 8583 CD, PG 8591, PG 8592, PG 8593
  • Cat. No.:
    141-0626
  • W×D×H:
    250×350×30 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.