Accessories
Connection module AM 10, PG 8535, PG 8536
Catalog # 141-0561
Supplier: MIELE
Jump to:
Product Details & Documents
- For lower and upper baskets
- With 7 x 7 mm perforations
- Stainless steel
Specifications
- Description:Connection module AM 10
- Seq:09
- For:PG 8535, PG 8536