Orders LinkContactUsLinkComponent
Inserts
Inserts
E 403 Insert suitable for 105 watch glasses Ø 50 - 60 mm, equipped with 36 bars spaced 9 mm apart
Catalog # 141-0388
Supplier:   MIELE
Inserts
Catalog # 141-0388
Supplier:  MIELE
Inserts
Inserts
E 403 Insert suitable for 105 watch glasses Ø 50 - 60 mm, equipped with 36 bars spaced 9 mm apart
Price
738.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    E 403 Insert suitable for 105 watch glasses Ø 50 - 60 mm, equipped with 36 bars spaced 9 mm apart
  • Seq:
    52