Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Inserts

Inserts

E 965 insert, with 15×3 holders for beakers 250–600 ml, G 7827, 357×522×173 mm
Catalog # 141-0470
Supplier:   MIELE
Inserts
Catalog # 141-0470
Supplier:  MIELE
Inserts
Inserts

E 965 insert, with 15×3 holders for beakers 250–600 ml, G 7827, 357×522×173 mm

Price
1 150.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    E 965 insert, with 15×3 holders for beakers 250–600 ml
  • Seq:
    23
  • W×D×H:
    357×522×173 mm
  • For:
    G 7827
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.