ContactUsLinkComponent
GBOX CHEMI XR5 camera, lens and bracket. CCD Mono camera 5 million pixels 65535 grey level dynamic range Peltier cooled. 1 * 1 items
Catalog # SYNO701-11055-01
GBOX CHEMI XR5 camera, lens and bracket. CCD Mono camera 5 million pixels 65535 grey level dynamic range Peltier cooled. 1 * 1 items
Catalog # SYNO701-11055-01
Frequently Bought Together