COD standard solutions
COD standard solution, volume: 30 ml, model: SL COD 400, 400 mg/l, 1
Supplier: WTW
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Concentration:400 mg/l
- Pk:1
- Description:COD standard solution, volume: 30 ml, model: SL COD 400
- Cat. No.:WTWA252451
- Physical form:Liquid



