COD standard solutions
COD standard solution, volume: 100 ml, model: SL COD 100, 100 mg/l, 1
Catalog # WTWA252450
Supplier: WTW
Jump to:
Product Details & Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Concentration:100 mg/l
- Pk:1
- Description:COD standard solution, volume: 100 ml, model: SL COD 100
- Cat. No.:WTWA252450
- Physical form:Liquid



