ContactUsLinkComponent
Hydrometer, ASTM 86H62, ISO 650, series L50, range 0.800-0.850, tolerance ± 0.0005, div 0,0005g/ml, calibration temperature 60/60 °F, with calibration certificate. Certificate of calibtration issued by our laboratory, using masters traceable to offic 1 * 1 items
Catalog # ALLA3510LC08060BEU
Supplier:  Alla France
Hydrometer, ASTM 86H62, ISO 650, series L50, range 0.800-0.850, tolerance ± 0.0005, div 0,0005g/ml, calibration temperature 60/60 °F, with calibration certificate. Certificate of calibtration issued by our laboratory, using masters traceable to offic 1 * 1 items
Catalog # ALLA3510LC08060BEU
Supplier:  Alla France
Frequently Bought Together