ContactUsLinkComponent
Hydrometer, ASTM 86H62, ISO 650, series L50, range 0.850-0.900, tolerance ± 0.0005, div 0,0005g/ml, calibration temperature 60/60 °F , with calibration certificate. 1 * 1 items
Catalog # ALLA3510LC08560BEU
Supplier:  Alla France
Hydrometer, ASTM 86H62, ISO 650, series L50, range 0.850-0.900, tolerance ± 0.0005, div 0,0005g/ml, calibration temperature 60/60 °F , with calibration certificate. 1 * 1 items
Catalog # ALLA3510LC08560BEU
Supplier:  Alla France
Frequently Bought Together