Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Laemmli SDS sample buffer, non-reducing (4X)

Laemmli SDS sample buffer, non-reducing (4X)

Laemmli SDS Sample Buffer, non-reducing, 4X, 50 mL
Catalog # J63615.AD
Laemmli SDS sample buffer, non-reducing (4X)
Catalog # J63615.AD
CAS Number:  
Laemmli SDS sample buffer, non-reducing (4X)
Laemmli SDS sample buffer, non-reducing (4X)

Laemmli SDS Sample Buffer, non-reducing, 4X, 50 mL

CAS Number:  
Price
SFr. 206.00
Log in for availability
1 * 50 mL
Product Details & Documents

Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system

Specifications

Specifications

  • Pk:
    50 mL
  • Description:
    Laemmli SDS Sample Buffer, non-reducing, 4X
  • Cat. No.:
    J63615.AD
  • Physical form:
    Liquid
  • Appearance:
    Blue

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.