IEF Anode buffer (50X)
IEF Anode buffer (50X), 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD03846400
Applications: For isoelectric focusing (IEF)
Documents
Specifications
Specifications
- Pk:500 mL
- Description:IEF Anode buffer (50X)
- Cat. No.:J63303.AP
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD03846400
- Appearance:Colourless
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



