Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Succinate, 0,2 M buffer solution, pH 5,0

Succinate, 0,2 M buffer solution, pH 5,0

Succinate, 0.2M buffer soln., pH 5.0, 500 mL
Catalog # J63863.AP
Succinate, 0,2 M buffer solution, pH 5,0
Catalog # J63863.AP
CAS Number:  
Succinate, 0,2 M buffer solution, pH 5,0
Succinate, 0,2 M buffer solution, pH 5,0

Succinate, 0.2M buffer soln., pH 5.0, 500 mL

CAS Number:  
Price
188.00 SFr.
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Succinate, 0.2M buffer soln., pH 5.0
  • Cat. No.:
    J63863.AP
  • Odour/scent:
    Odourless
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00002789
  • Appearance:
    Colourless
  • CAS No.:
    150-90-3

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.