Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Succinate, 0,2 M buffer solution, pH 5,0

Succinate, 0.2M buffer soln., pH 5.0, 250 mL
  J63863.AK
Succinate, 0,2 M buffer solution, pH 5,0
  J63863.AK
 :  
Succinate, 0,2 M buffer solution, pH 5,0
Succinate, 0,2 M buffer solution, pH 5,0

Succinate, 0.2M buffer soln., pH 5.0, 250 mL

 :  
 
119.00 SFr.
   
1 * 250 mL
 

 

  • Pk:
    250 mL
  • Description:
    Succinate, 0.2M buffer soln., pH 5.0
  • Cat. No.:
    J63863.AK
  • Odour/scent:
    Odourless
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00002789
  • Appearance:
    Colourless
  • CAS No.:
    150-90-3