Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Methotrexate

Methotrexate

Glass bottle for solids, 100 mg
Catalog # J63075.MC
Toxic
Health
Irritant
Methotrexate
Catalog # J63075.MC
CAS Number:  59-05-2
Methotrexate
Methotrexate

Glass bottle for solids, 100 mg

CAS Number:  59-05-2
Price
SFr. 275.00
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
  • CAS Number:59-05-2
  • Formula:C₂₀H₂₂N₈O₅
  • MDL Number:MFCD00150847
  • Molecular Weight:454.44 g/mol
  • Merck Index:14,5985
  • Storage Temperature:Freezer
  • EINECS:200-413-8
  • UN:1544
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.