α-Cyclodextrin ≥97%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 97+% CAS No.: 10016-20-3 Molecular Formula: C36H60O30 Formula Weight: 972.84 Physical Form: Powder Appearance: White Solubility: Soluble in water at 1%(w/v) Merck Reference: 14,2718 Optical Rotation: +136° (c=10 in water) MDL No.: MFCD00078207
Useful for selective precipitation of enantiomeric, positional or structural isomers
- CAS Number:10016-20-3
- Formula:C₃₆H₆₀O₃₀
- MDL Number:MFCD00078207
- Molecular Weight:972.84 g/mol
- Merck Index:14,2718
- Storage Temperature:Ambient
- EINECS:233-007-4
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



