Order Entry
Switzerland
Orders LinkContactUsLinkComponent
α-Cyclodextrin ≥97%

α-Cyclodextrin ≥97%

Glass bottle for solids, 5 g
Catalog # J60687.06
α-Cyclodextrin ≥97%
Catalog # J60687.06
CAS Number:  10016-20-3
α-Cyclodextrin ≥97%
α-Cyclodextrin ≥97%

Glass bottle for solids, 5 g

CAS Number:  10016-20-3
Price
103.00 SFr.
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 97+% CAS No.: 10016-20-3 Molecular Formula: C36H60O30 Formula Weight: 972.84 Physical Form: Powder Appearance: White Solubility: Soluble in water at 1%(w/v) Merck Reference: 14,2718 Optical Rotation: +136° (c=10 in water) MDL No.: MFCD00078207

Useful for selective precipitation of enantiomeric, positional or structural isomers

  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  • Merck Index:14,2718
  • Storage Temperature:Ambient
  • EINECS:233-007-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications